missing translation for 'onlineSavingsMsg'
Learn More

Chlorocarbonylsulfenyl chloride, 95%

Catalog No. AC224340050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224340050 5 g Glass bottle
AC224340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224340050 Supplier Thermo Scientific Chemicals Supplier No. 224340050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2757-23-5 | CCl2OS | 130.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2757-23-5
Molecular Formula CCl2OS
Molecular Weight (g/mol) 130.97
MDL Number MFCD00000703
InChI Key MNOALXGAYUJNKX-UHFFFAOYSA-N
Synonym chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
PubChem CID 75990
SMILES ClSC(Cl)=O

Specifications

Color Yellow
Density 1.5800g/mL
Boiling Point 98°C to 100°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClCOSCl
Quantity 5 g
Specific Gravity 1.58
Solubility Information Solubility in water: reacts
IUPAC Name chloro(chlorosulfanyl)methanone
Formula Weight 130.98
Percent Purity 95%
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Chlorocarbonylsulfenyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-415-2

RUO – Research Use Only