missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobutanol, Hydrous, NF, 98-100.5%, Spectrum™ Chemical
CH122, 6001-64-5, C4H7Cl3O·1/2H2O
Supplier: Spectrum Chemical Mfg Cor CH12250KGBL
Description
Spectrum™ Chemical Chlorobutanol, Hydrous, NF is a very effective preservative in many pharmaceutical products for eyes, ears and nose, dental preparations. The hemihydrate form is useful in formulations where the absence of water is not critical. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 6001-64-5 | |
| Fiber Drum | |
| 50 kg | |
| O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl | |
| 372.91 | |
| NF |
| 100% | |
| C8H16Cl6O3 | |
| WRWLCXJYIMRJIN-UHFFFAOYSA-N | |
| bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate | |
| 98-100.5% |
Chemical Identifiers
| 6001-64-5 | |
| 372.91 | |
| bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate |
| C8H16Cl6O3 | |
| WRWLCXJYIMRJIN-UHFFFAOYSA-N | |
| O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl |