missing translation for 'onlineSavingsMsg'
Learn More

Chlorobutanol, Hydrous, NF, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7157261
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
125 g
2.5 kg
25 g
50 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18601605 25 g Amber Glass Bottle
18601606 125 g Amber Glass Bottle
18601607 500 g Amber Glass Bottle
18601610 2.5 kg Poly Pail
18601608 12 kg Poly Pail
18601609 50 kg Fiber Drum
6 options

Catalog No. 18601605

Supplier: Spectrum Chemical Mfg Cor CH12225GM

Only null left

CH122, 6001-64-5, C4H7Cl31/2H2O

Spectrum™ Chemical Chlorobutanol, Hydrous, NF is a very effective preservative in many pharmaceutical products for eyes, ears and nose, dental preparations. The hemihydrate form is useful in formulations where the absence of water is not critical. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6001-64-5
Molecular Formula C8H16Cl6O3
Molecular Weight (g/mol) 372.91
InChI Key WRWLCXJYIMRJIN-UHFFFAOYSA-N
IUPAC Name bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate
SMILES O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98-100.5%
Grade NF