missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(ethylene)rhodium(I) Dimer 95.0+%, TCI America™

Catalog No. C2461200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2461200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2461200MG Supplier TCI America Supplier No. C2461200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Molecular Weight (g/mol) 388.93
MDL Number MFCD00013206
InChI Key OOQJCPOXJFCGCR-UHFFFAOYSA-L
Synonym chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i
PubChem CID 45357812
IUPAC Name bis(chlororhodium); tetrakis(ethene)
SMILES Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C

Specifications

Quantity 200 mg
Formula Weight 388.93
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorobis(ethylene)rhodium(I) Dimer
TSCA No
Recommended Storage Freezer (-20 °C)