missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(ethylene)iridium(I) Dimer 95.0+%, TCI America™

Catalog No. c3041100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3041100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3041100MG Supplier TCI America Supplier No. C3041100MG
Only null left

Chemical Identifiers

CAS 39722-81-1
Molecular Formula C8H16Cl2Ir2-2
Molecular Weight (g/mol) 567.55
InChI Key KIGDBTYHWLDKQG-UHFFFAOYSA-L
Synonym Dichlorotetra(ethylene)diiridium(I)
PubChem CID 91972112
IUPAC Name ethene;iridium;dichloride
SMILES C=C.C=C.C=C.C=C.[Cl-].[Cl-].[Ir].[Ir]

Specifications

Quantity 100 mg
Formula Weight 567.55
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorobis(ethylene)iridium(I) Dimer
TSCA No
Recommended Storage Refrigerator