accessibility menu, dialog, popup

UserName

CH2H2CCH2H2CCH2H2CCH2H2CClClIrIr

CAS RN 39722-81-1

IUPAC Name: tetrakis(ethene) diiridium dichloride
Synonyms: tetrakis(ethylene) diiridium dichloride
Molecular Weight (g/mol): 567.55
Molecular Formula: C8H16Cl2Ir2
InChi Key: KIGDBTYHWLDKQG-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ir].[Ir].C=C.C=C.C=C.C=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results