missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, ACS Reagent (Monochlorobenzene), Reagents
Chlorobenzene, ACS Reagent (Monochlorobenzene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2175100500C
Specifications
| Chlorobenzene, ACS Reagent (Monochlorobenzene) | |
| 108-90-7 | |
| 132°C | |
| Glass Bottle | |
| 1.107 | |
| Monochlorobenzene, Phenyl chloride, Benzene chloride | |
| ClC1=CC=CC=C1 | |
| 112.56 |
| -45°C | |
| Colorless | |
| 500 mL | |
| Liquid | |
| C6H5Cl | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| chlorobenzene | |
| ACS |
Chemical Identifiers
| 108-90-7 | |
| 112.56 | |
| Monochlorobenzene, Phenyl chloride, Benzene chloride | |
| ClC1=CC=CC=C1 |
| C6H5Cl | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| chlorobenzene |