missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ACS Reagent (Monochlorobenzene), Reagents

Catalog No. p-200020941
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337325 4 L
01337324 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337325 Supplier Labchem Inc Supplier No. C21751004C
Only null left

Chlorobenzene, ACS Reagent (Monochlorobenzene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.56
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym Monochlorobenzene, Phenyl chloride, Benzene chloride
IUPAC Name chlorobenzene
SMILES ClC1=CC=CC=C1

Specifications

Melting Point -45°C
Color Colorless
Boiling Point 132°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.107
Grade ACS
Chemical Name or Material Chlorobenzene, ACS Reagent (Monochlorobenzene)