missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil

Catalog No. p-7081775
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 g
250 g
Catalog No. Quantity
AAA1192230 250 g
AAA119220B 1000 g
2 options

Catalog No. AAA1192230

Supplier: Thermo Scientific Chemicals A1192230

Only null left

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

Solubility
Soluble in water (124g/L), ether and chloroform.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES CC(=O)CCl

Specifications

Melting Point °C to 45°C
Density 1.161
Boiling Point 118°C to 120°C
Flash Point 40°C (104°F)
Assay Percent Range 95%, Stabilized with 0.1% Epoxidized Soybean Oil
Linear Formula CH2COCH3
Refractive Index 1.434
Quantity 250 g
UN Number UN1695
Beilstein 605369
Merck Index 14,2114
Synonym 1-propanol, 3-chloro-, acetate, 3-chloropropanol acetate, 3-chlorprop-1-ylacetat, gamma-chloropropyl acetate, 3-chlorprop-1-ylacetat german, acetic acid 3-chloro-propyl ester, 1-acetoxy-3-chloropropane, .gamma.-chloropropyl acetate, 3-chloro-propyl acetate, acetic acid 3-chloropropyl ester
Solubility Information Soluble in water (124g/L),ether and chloroform.
IUPAC Name 3-chloropropyl acetate
PubChem CID 12334
Formula Weight 92.53
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized with 0.1% epoxidized soybean oil
Show More Show Less
Hazard Category H226-H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H310-H330-H314-H318-H226
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETONE, STABILIZED
EINECSNumber 201-161-1
RTECSNumber UC0700000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only