accessibility menu, dialog, popup

UserName

CAS RN 78-95-5

IUPAC Name: 1-chloropropan-2-one
Synonyms: tonite α-stoff chloroacetone (substance)
Molecular Weight (g/mol): 92.52
Molecular Formula: C3H5ClO
InChi Key: BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES: CC(=O)CCl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)