missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 391610010

 View more versions of this product

Catalog No. AC391610010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Chloro(triphenylphosphine)gold(I)
Authentic
Glass bottle
MFCD00009588
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
IFPWCRBNZXUWGC-UHFFFAOYSA-M
chlorogold;triphenylphosphane
10874691
99%
14243-64-2
98.5% min. (ex Au)
C18H15AuClP
1g
Solubility in water: insoluble
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Au]
494.71
494.71

Chemical Identifiers

14243-64-2
494.71
IFPWCRBNZXUWGC-UHFFFAOYSA-M
10874691
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Au]
C18H15AuClP
MFCD00009588
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
chlorogold;triphenylphosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 238-117-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only