missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate

Catalog No. AC439070010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439070010 1 g
AC439070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439070010 Supplier Thermo Scientific Chemicals Supplier No. 439070010

CAS: 207915-99-9 | C5H12ClF6N2P | 280.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 207915-99-9
Molecular Formula C5H12ClF6N2P
Molecular Weight (g/mol) 280.58
MDL Number MFCD01862891
InChI Key CUKNPSDEURGZCO-UHFFFAOYSA-N
Synonym chloro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, tcfh, n-chloro dimethylamino methylene-n-methylmethanaminium hexafluorophosphate v, n,n,n',n'-tetramethylchloroformamidinium hexafluorophosphate, tcfh, chloro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n,n,n,n-tetramethylchloroformamidinium hexafluorophosphate, chloro dimethylamino methylidene dimethylazanium hexafluorophosphate, pubchem12759, acmc-209rtw, c5h12cln2.f6p
PubChem CID 10989639
IUPAC Name [chloro(dimethylamino)methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CN(C)C(Cl)=[N+](C)C

Specifications

Melting Point 102.4°C to 104.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 1 g
Formula Weight 280.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only