accessibility menu, dialog, popup

UserName

CAS RN 207915-99-9

IUPAC Name: [chloro(dimethylamino)methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Synonyms: [chloro(dimethylamino)methylidene]dimethylazanium hexafluorophosphate
Molecular Weight (g/mol): 280.58
Molecular Formula: C5H12ClF6N2P
InChi Key: CUKNPSDEURGZCO-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)C(Cl)=[N+](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results