missing translation for 'onlineSavingsMsg'
Learn More

Chloro(methyl)(phenyl)(vinyl)silane 98.0+%, TCI America™

Catalog No. C34345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34345G 5 g
C343425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34345G Supplier TCI America Supplier No. C34345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17306-05-7
Molecular Formula C9H11ClSi
Molecular Weight (g/mol) 182.722
MDL Number MFCD00053208
InChI Key GSXJAPJSIVGONK-UHFFFAOYSA-N
PubChem CID 87042
IUPAC Name chloro-ethenyl-methyl-phenylsilane
SMILES C[Si](C=C)(C1=CC=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 2987
Formula Weight 182.72
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(methyl)(phenyl)(vinyl)silane
EINECSNumber (3)-1165
TSCA Yes