missing translation for 'onlineSavingsMsg'
Learn More

Chloro(ethyl)dimethylsilane 97.0+%, TCI America™

Catalog No. D01355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01355G 5 g
D013525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01355G Supplier TCI America Supplier No. D01355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6917-76-6
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00009918
InChI Key AVDUEHWPPXIAEB-UHFFFAOYSA-N
Synonym ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
PubChem CID 138863
IUPAC Name chloro-ethyl-dimethylsilane
SMILES CC[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 2985
Formula Weight 122.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(ethyl)dimethylsilane [Dimethylethylsilylating Agent]
EINECSNumber (2)-2041
TSCA No