CAS RN 6917-76-6
IUPAC Name:
chloro(ethyl)dimethylsilane
Synonyms:
chloro(ethyl)dimethylsilane
Molecular Weight (g/mol):
122.67
Molecular Formula:
C4H11ClSi
InChi Key:
AVDUEHWPPXIAEB-UHFFFAOYSA-N
SMILES:
CC[Si](C)(C)Cl
1
–
1
of
1
results
1
Chloro(ethyl)dimethylsilane 97.0+%, TCI America™
CAS: 6917-76-6 Molecular Formula: C4H11ClSi Molecular Weight (g/mol): 122.667 MDL Number: MFCD00009918 InChI Key: AVDUEHWPPXIAEB-UHFFFAOYSA-N Synonym: ethyldimethylchlorosilane,chlorodimethylethylsilane,chloro ethyl dimethylsilane,dimethylethylchlorosilane,dimethylethylsilyl chloride,ethyldimethylsilyl chloride,chloro ethyl dimethylsilane #,silane, chlorodimethylethyl-, PubChem CID: 138863 IUPAC Name: chloro-ethyl-dimethylsilane SMILES: CC[Si](C)(C)Cl