missing translation for 'onlineSavingsMsg'
Learn More

Chloro(dimethylsulfide)gold(I) 97.0+%, TCI America™

Catalog No. C2719200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2719200MG 200 mg
C27191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2719200MG Supplier TCI America Supplier No. C2719200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29892-37-3
Molecular Formula C2H6AuClS
Molecular Weight (g/mol) 294.547
MDL Number MFCD00192090
InChI Key YQALRAGCVWJXGB-UHFFFAOYSA-M
Synonym (Dimethylsulfide)gold(I) Chloride
PubChem CID 6100873
IUPAC Name chlorogold;methylsulfanylmethane
SMILES CSC.Cl[Au]

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 200 mg
Formula Weight 294.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro(dimethylsulfide)gold(I)
TSCA No
Recommended Storage Refrigerator