missing translation for 'onlineSavingsMsg'
Learn More

Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals

Catalog No. AA4663903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4663903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4663903 Supplier Thermo Scientific Chemicals Supplier No. 04663903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II) acts as a ligand and chiral catalyst in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1252598-33-6
Molecular Formula C52H54ClFePPd
Molecular Weight (g/mol) 907.69
MDL Number MFCD25372545
InChI Key ZDJQQIXNGDRDQM-UHFFFAOYSA-M
Synonym pdcl crotyl qphos, chloro crotyl 1,2,3,4,5-pentaphenyl-1'-di-tert-butylphosphino ferrocene palladium ii
PubChem CID 73994979
IUPAC Name λ²-iron(2+) 3-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide but-2-en-1-ide chloropalladiumylium pentaphenylcyclopenta-2,4-dien-1-ide
SMILES Cl[Pd++].C\C=C/C.CC(C)(C)P(c1cccc1[Fe])C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Pink
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 907.69
Physical Form Solid
Chemical Name or Material Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only