accessibility menu, dialog, popup

UserName

FePPdCH4H3CCH3ClPhPhPhPhPht-But-Bu

CAS RN 1252598-33-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 907.69
Molecular Formula: C52H54ClFePPd
InChi Key: —
SMILES: Cl[Pd++].C\C=C/C.CC(C)(C)P(c1cccc1[Fe])C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results