missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(chloromethyl)dimethylsilane 98.0+%, TCI America™
Supplier: TCI America C0605250G
Specifications
| Chloro(chloromethyl)dimethylsilane | |
| 115°C | |
| MFCD00000875 | |
| 2985 | |
| ITKVLPYNJQOCPW-UHFFFAOYSA-N | |
| chloro(chloromethyl)dimethylsilane | |
| 74393 | |
| ≥98.0% (GC) |
| 1719-57-9 | |
| C3H8Cl2Si | |
| 250 g | |
| chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane | |
| C[Si](C)(Cl)CCl | |
| 143.08 | |
| 143.08 | |
| Liquid |
Chemical Identifiers
| 1719-57-9 | |
| 143.08 | |
| ITKVLPYNJQOCPW-UHFFFAOYSA-N | |
| 74393 | |
| C[Si](C)(Cl)CCl |
| C3H8Cl2Si | |
| MFCD00000875 | |
| chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane | |
| chloro(chloromethyl)dimethylsilane |
Safety and Handling
EINECSNumber : (2)-2047
TSCA : Yes