missing translation for 'onlineSavingsMsg'
Learn More

Chloro(chloromethyl)dimethylsilane 98.0+%, TCI America™

Catalog No. p-7124521
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060525G 25 g
C0605250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C060525G Supplier TCI America Supplier No. C060525G

Chemical Identifiers

CAS 1719-57-9
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.08
MDL Number MFCD00000875
InChI Key ITKVLPYNJQOCPW-UHFFFAOYSA-N
Synonym chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
PubChem CID 74393
IUPAC Name chloro(chloromethyl)dimethylsilane
SMILES C[Si](C)(Cl)CCl

Specifications

Boiling Point 115°C
Quantity 25 g
UN Number 2985
Formula Weight 143.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(chloromethyl)dimethylsilane
EINECSNumber (2)-2047
TSCA Yes