missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) Dimer 98.0+%, TCI America™

Catalog No. p-7118823
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1045100MG 100 mg
B10451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1045100MG Supplier TCI America Supplier No. B1045100MG
Only null left

Chemical Identifiers

CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Molecular Weight (g/mol) 493.08
MDL Number MFCD00012415
InChI Key QSUDXYGZLAJAQU-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer
PubChem CID 6436379
IUPAC Name bis(λ¹-rhodium(1+)) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Quantity 100 mg
Formula Weight 493.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1,5-cyclooctadiene)rhodium(I) Dimer