missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh

Catalog No. AC211305000
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211305000 500 mg Glass bottle
AC211300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211305000 Supplier Thermo Scientific Chemicals Supplier No. 211305000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12092-47-6 | C16H24Cl2Rh2 | 493.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Molecular Weight (g/mol) 493.08
MDL Number MFCD00012415
InChI Key QSUDXYGZLAJAQU-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer
PubChem CID 6436379
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 256.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: 0.34g/L
Formula Weight 493.08
Percent Purity ≥40.8% (RH)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Chloro(1, 5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 235-157-6
TSCA TSCA

RUO – Research Use Only