missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) 98.0+%, TCI America™
$109.98 - $298.25
Chemical Identifiers
| CAS | 578743-87-0 |
|---|---|
| Molecular Formula | C27H36ClCuN2 |
| MDL Number | MFCD09264276 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2304200MG
|
TCI America
C2304200MG |
200 mg |
Each for $109.98
|
|
|||||
|
C23041G
|
TCI America
C23041G |
1 g |
Each for $298.25
|
|
|||||
Chemical Identifiers
| 578743-87-0 | |
| C27H36ClCuN2 | |
| MFCD09264276 |
Specifications
| 578743-87-0 | |
| C27H36ClCuN2 | |
| 200 mg | |
| ≥98.0% (T) | |
| Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) |
| White-Yellow | |
| MFCD09264276 | |
| 487.60 | |
| Crystalline Powder |
Safety and Handling
TSCA : No