missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorite IC Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| CAS | 7758-19-2 |
| Concentration | 1000 μg/mL in H 2 O |
| Type | Analytical Standard |
| Quantity | 100 mL |
| Grade | Analytical Standard |