missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorine in Heavy Mineral Oil standard solution, Specpure™ blank (0.000%)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Chlorine in Heavy Mineral Oil standard solution, Blank (0.000%) |
Safety and Handling
| Hazard Category | H304 |
| Hazard Statement | GHS H Statement H350 May cause cancer. |
| Precautionary Statement | P301+P310-P331-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only