missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ASCL95X

 View more versions of this product

Catalog No. AS-CL9-5X

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cl&superscript_minus;0.1M

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Chloride Standard
With Certificate of Analysis
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011420
Certified Reference Material
N.Cl
azanium;chloride
500 mL
CHEBI:31206
Triple-leached LDPE bottle
12125-02-9,7732-18-5
ClH4N
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
NLXLAEXVIDQMFP-UHFFFAOYSA-N
Water
53.49
25517
0.1M

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
CHEBI:31206
ClH4N
MFCD00011420
25517
azanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Non-Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only