missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic Acid Sodium Salt Hydrate 95.0+%, TCI America™

Catalog No. c00815g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00815G 5 g
C008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00815G Supplier TCI America Supplier No. C00815G
Only null left

Chemical Identifiers

CAS 36275-66-8
Molecular Formula C6Cl2Na2O4
Molecular Weight (g/mol) 252.942
MDL Number MFCD00013787
InChI Key FITANBHHCLWRKG-UHFFFAOYSA-L
Synonym chloranilic acid sodium salt, chloranilic acid disodium salt, disodium chloranilate, chloroanilic acid sodium salt, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, disodium salt, sodium chloranilate, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, sodium salt 1:2, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate, chloranilic acid sodium salt hydrate, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis olate
PubChem CID 118938
IUPAC Name disodium;2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)[O-])Cl)[O-].[Na+].[Na+]

Specifications

Quantity 5 g
Formula Weight 252.94
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloranilic Acid Sodium Salt Hydrate