accessibility menu, dialog, popup

UserName

NaNaOClOClOO

CAS RN 36275-66-8

IUPAC Name: disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate)
Synonyms: disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate)
Molecular Weight (g/mol): 252.94
Molecular Formula: C6Cl2Na2O4
InChi Key: FITANBHHCLWRKG-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=C(Cl)C(=O)C([O-])=C(Cl)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results