missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloranilic Acid Lanthanum(III) Salt Decahydrate 98.0+%, TCI America™
Supplier: TCI America C007925G
Specifications
| Chloranilic Acid Lanthanum(III) Salt Decahydrate | |
| 32607-23-1 | |
| 25 g | |
| MFCD00135764 | |
| GGKWUCDKAJOTCW-UHFFFAOYSA-N | |
| 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;lanthanum | |
| 126959841 | |
| ≥98.0% (T) |
| Black | |
| Crystalline Powder | |
| C18H6Cl6La2O12 | |
| chloranilic acid lanthanum salt, chloranilic acid, lanthanum salt | |
| C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.[La].[La] | |
| 904.745 | |
| 898.70 |
Chemical Identifiers
| 32607-23-1 | |
| 904.745 | |
| GGKWUCDKAJOTCW-UHFFFAOYSA-N | |
| 126959841 | |
| C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.[La].[La] |
| C18H6Cl6La2O12 | |
| MFCD00135764 | |
| chloranilic acid lanthanum salt, chloranilic acid, lanthanum salt | |
| 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;lanthanum |
Safety and Handling
TSCA : Yes