missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chloramphenicol, 98%

Catalog No. AC227920250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227920250 25 g Glass Bottle
AC227921000 100 g Glass Bottle
AC227925000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227920250 Supplier Thermo Scientific Chemicals Supplier No. 227920250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chloramphenicol, CAS # 56-75-7, is a preparation of the broad-spectrum, semisynthetic antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Chloramphenicol is an organochloride compound that contains a nitrobenzene ring
  • Chloramphenicol acts as a bacteriostatic by binding the 50S ribosomal subunit and preventing peptide formation
  • Stops peptidyltransferase activity by complexing to 50S subunit

Applications

  • Intended for molecular and microbiology applications such as antibiotic resistance gene testing and selective growth experiments and as a selection agent in bacterial cell culture
  • For research use only

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
MDL Number MFCD00078159
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
Synonym Chloromycetin, D(-)-threo-2, 2-Dichloro-N-[β-hydroxy-α-(hydroxy-methyl)-p-nitrophenylethyl]acetamide
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Type Chloramphenicol
Melting Point 149.0°C to 153.0°C
Packaging Glass Bottle
Specific Rotation + 18.50
Quantity 25 g
Assay Percent Range 98%
Solubility Information Solubility in water: 2.5g/L (25°C). Other solubilities: 150.8 mg/mL propylene glycol, soluble in methanol, ethanol, butanol, acetone and, ethyl acetate - slightly soluble in ether, insoluble in benzene and petroleum ether
Formula Weight 323.13
Percent Purity 98%
Chemical Name or Material Chloramphenicol
Hazard Statement GHS H Statement:
May cause cancer.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only