missing translation for 'onlineSavingsMsg'
Learn More

Chloramine T Trihydrate 98.0+%, TCI America™

Catalog No. p-7124205
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C007625G 25 g
C0076500G 500 g
2 options

Catalog No. C007625G

Supplier: TCI America C007625G

Only null left

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
Synonym benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
PubChem CID 122197679
IUPAC Name sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3263
Formula Weight 227.64
Percent Purity ≥98.0% (T)
Chemical Name or Material Chloramine T Trihydrate
EINECSNumber (3)-2178&(3)-3132
RTECSNumber XT5616800
TSCA Yes