missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramine-T Trihydrate, Reagent, ACS, 98-103%, Spectrum™ Chemical
CH120, 7080-50-4, C7H7SO2NClNa·3H2O
$237.31 - $1663.77
Chemical Identifiers
| Molecular Formula | C7H13ClNNaO5S |
|---|---|
| Molecular Weight (g/mol) | 281.68 |
| MDL Number | MFCD00149066 |
| InChI Key | RNROOTWAJBXJBD-UHFFFAOYSA-M |
| IUPAC Name | sodium (chloroimino)(4-methylphenyl)-λâ¶-sulfanoylolate trihydrate |
| SMILES | O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601581
|
Spectrum Chemical Mfg Cor
CH120125GM |
125 g | Amber Glass Bottle |
Each for $237.31
|
|
||||
|
18601585
|
Spectrum Chemical Mfg Cor
CH120250GM |
250 g | Amber Glass Bottle |
Each for $342.17
|
|
||||
|
18601582
|
Spectrum Chemical Mfg Cor
CH120500GM |
500 g | Amber Glass Bottle |
Each for $454.80
|
|
||||
|
18601583
|
Spectrum Chemical Mfg Cor
CH12025KG |
2.5 kg | Poly Pail |
Each for $1,663.77
|
|
||||
Description
Spectrum™ Chemical Chloramine-T Trihydrate, Reagent, ACS is also known as tosylchloramide. It is most commonly used as a disinfectant or as a biocide. It is a white powder and can be a source of electrophilic chlorine in organic synthesis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| C7H13ClNNaO5S | |
| MFCD00149066 | |
| sodium (chloroimino)(4-methylphenyl)-λâ¶-sulfanoylolate trihydrate |
| 281.68 | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
Specifications
| 100% | |
| C7H13ClNNaO5S | |
| UN3263 | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl | |
| 281.68 | |
| ACS |
| 125 g | |
| MFCD00149066 | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| sodium (chloroimino)(4-methylphenyl)-λâ¶-sulfanoylolate trihydrate | |
| 98 to 103% | |
| Amber Glass Bottle |