missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T Trihydrate, Reagent, ACS, 98-103%, Spectrum™ Chemical

Catalog No. 18601581
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601581 125 g Amber Glass Bottle
18601585 250 g Amber Glass Bottle
18601582 500 g Amber Glass Bottle
18601583 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601581 Supplier Spectrum Chemical Mfg Cor Supplier No. CH120125GM

CH120, 7080-50-4, C7H7SO2NClNa·3H2O

Spectrum™ Chemical Chloramine-T Trihydrate, Reagent, ACS is also known as tosylchloramide. It is most commonly used as a disinfectant or as a biocide. It is a white powder and can be a source of electrophilic chlorine in organic synthesis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
IUPAC Name sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

CAS Max % 100%
Quantity 125 g
UN Number UN3263
Percent Purity 98 to 103%
Grade ACS
Packaging Amber Glass Bottle