missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, ACS, 98.0-103.0%

Catalog No. AA4237436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4237436 500 g
AA4237422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4237436 Supplier Thermo Scientific Chemicals Supplier No. 04237436

Reagent for selective oxidation of methionine | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
Synonym benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
PubChem CID 122197679
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Melting Point 167°C to 170°C
Color White to Yellow
pH 8 to 10
Physical Form Crystalline
Quantity 500 g
UN Number UN3263
Beilstein 3924168
Flash Point 192°C (377°F)
Merck Index 14,2075
Solubility Information Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Formula Weight 281.69 (227.67 Anhydrous)
Percent Purity 98.0 to 103.0%
Grade ACS
Odor Chlorine-like
Sensitivity Moisture sensitive
Chemical Name or Material Chloramine-T trihydrate
Show More Show Less
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H334-H314-H318-H302
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 204-854-7
RTECSNumber XT5616800
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only