missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Chlorambucil | |
| 67°C | |
| C14H19Cl2NO2 | |
| 1 g | |
| chlorambucil, ambochlorin, chloroambucil, leukeran, chloraminophen, chlorbutin, chloraminophene, chlorobutine, amboclorin, chlorbutine | |
| OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl | |
| 304.21 | |
| CHEBI:28830 | |
| ≥98.0% (GC,T) |
| 305-03-3 | |
| White-Yellow | |
| MFCD00021783 | |
| 2811 | |
| JCKYGMPEJWAADB-UHFFFAOYSA-N | |
| 4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid | |
| 2708 | |
| 304.21 | |
| Crystalline Powder |
Chemical Identifiers
| 305-03-3 | |
| 304.21 | |
| JCKYGMPEJWAADB-UHFFFAOYSA-N | |
| 2708 | |
| 4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid |
| C14H19Cl2NO2 | |
| MFCD00021783 | |
| chlorambucil, ambochlorin, chloroambucil, leukeran, chloraminophen, chlorbutin, chloraminophene, chlorobutine, amboclorin, chlorbutine | |
| CHEBI:28830 | |
| OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl |
Safety and Handling
RTECSNumber : ES7525000
TSCA : No
Recommended Storage : Refrigerator