accessibility menu, dialog, popup

UserName

CAS RN 305-03-3

IUPAC Name: 4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid
Synonyms: chlorambucil chlorbutine chlorobutin phenylbutyric acid nitrogen mustard elcoril elcorin ambochlorin chloorambucol 苯丁酸氮芥 glaxosmithkline brand of chlorambucil
Molecular Weight (g/mol): 304.21
Molecular Formula: C14H19Cl2NO2
InChi Key: JCKYGMPEJWAADB-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results