missing translation for 'onlineSavingsMsg'
Learn More

Chlorambucil 98.0+%, TCI America™

Catalog No. C2939200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2939200MG 200 mg
C29391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2939200MG Supplier TCI America Supplier No. C2939200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 305-03-3
Molecular Formula C14H19Cl2NO2
Molecular Weight (g/mol) 304.21
MDL Number MFCD00021783
InChI Key JCKYGMPEJWAADB-UHFFFAOYSA-N
Synonym chlorambucil, ambochlorin, chloroambucil, leukeran, chloraminophen, chlorbutin, chloraminophene, chlorobutine, amboclorin, chlorbutine
PubChem CID 2708
ChEBI CHEBI:28830
IUPAC Name 4-{4-[bis(2-chloroethyl)amino]phenyl}butanoic acid
SMILES OC(=O)CCCC1=CC=C(C=C1)N(CCCl)CCCl

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 304.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorambucil
RTECSNumber ES7525000
TSCA No
Recommended Storage Refrigerator