missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149949
Description
CHL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.Specifications
| CHL1 | |
| Polyclonal | |
| Unconjugated | |
| CHL1 | |
| A530023M13Rik; AI465420; CALL; cell adhesion molecule L1 like; cell adhesion molecule L1-like; cell adhesion molecule with homology to L1CAM; cell adhesion molecule with homology to L1CAM (close homolog of L1); cell adhesion molecule with homology to L1CAM (close homologue of L1); Chl1; Chl1-like protein; close homolog of L1; L1 cell adhesion molecule 2; L1CAM2; LICAM2; neural cell adhesion molecule L1-like protein; Processed neural cell adhesion molecule L1-like protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10752 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O00533 | |
| CHL1 | |
| Recombinant Cell Adhesion Molecule With Homology To L1CAM (CHL1) (Pro35-Glu328). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |