missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6235 Standard Phase Change Cooling Vest

Chill-Its™ 6235 Standard Phase Change Cooling Vest keeps workers cool and safe when combined with a cooling pack.

$121.60 - $121.60

Specifications

Material Cotton
Product Type Cooling Vest
Color Tan
For Use With (Application) Fire, Rescue, EMT, Construction, Utilities
Quantity 6/Pk.
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-819-666
View Documents
Ergodyne™
12004
Large/X-Large
Each for $121.60
Only null left
 
Description

Description

  • Adjustable hook and loop closures on both shoulders and waist
  • Constructed of cotton
  • Extended vest surface area enhances cooling power
Specifications

Specifications

Cotton
Tan
6/Pk.
Cooling Vest
Fire, Rescue, EMT, Construction, Utilities
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'