missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6235 Standard Phase Change Cooling Vest

Catalog No. 19819666
Change view
Click to view available options
Size:
Large/X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19819666 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819666 Supplier Ergodyne™ Supplier No. 12004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6235 Standard Phase Change Cooling Vest keeps workers cool and safe when combined with a cooling pack.

  • Adjustable hook and loop closures on both shoulders and waist
  • Constructed of cotton
  • Extended vest surface area enhances cooling power

Specifications

Material Cotton
Product Type Cooling Vest
Size Large/X-Large
Length (Metric) 102 to 132 cm
Length (English) 40 to 52 in.
Color Tan
For Use With (Application) Fire, Rescue, EMT, Construction, Utilities
Quantity 6/Pk.