missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594

Catalog No. A21471
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21471 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21471 Supplier Invitrogen™ Supplier No. A21471
  • ONLINE EXCLUSIVE PRICE
    $520.00
    $390.00 / Each

    Save $130.00

    25.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

Chicken anti-rat antibodies react with IgG heavy chains and all classes of immunoglobulin light chains from rat. Chicken secondary antibodies have gained popularity because they demonstrate a lower level of nonspecific binding. Chicken antibodies lack a classic 'Fc' domain and will not bind to protein A or protein G, nor will they bind to mammalian IgG Fc receptors. Product will be shipped at Room Temperature.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgG (H+L) Cross-Adsorbed
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 594
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Chicken
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less