missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488

Numéro de catalogue. A21441
Encompass
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A21441 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A21441 Fournisseur Invitrogen™ Code fournisseur A21441
  • ONLINE EXCLUSIVE PRICE
    $510.00
    $382.50 / chaque

    Économisez $127.50

    25.0 % de rabais Les écarts et divulgations suivants peuvent entraîner des prix inférieurs aux rabais indiqués, sans toutefois entraîner de réductions de prix supplémentaires : Le prix affiché reflète une réduction promotionnelle temporaire. Aucun achat additionnel ni engagement commercial n’est requis pour bénéficier du prix réduit. Le prix peut être modifié en tout temps. Le prix promotionnel est valide uniquement pour les commandes en ligne. Votre tarification contractuelle peut différer. D’autres exclusions peuvent s’appliquer.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Chicken Polyclonal Secondary Antibody

To minimize cross-reactivity, these chicken anti-rabbit IgG (H+L) whole secondary antibodies have been affinity purified and cross-adsorbed against human and mouse IgG prior to conjugation. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there are may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 488 dye is a bright, green-fluorescent dye with excitation ideally suited to the 488 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 488 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 488 dye molecules can be attached to proteins at high molar ratios without sig...

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Rabbit IgG (H+L) Cross-Adsorbed
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugué Alexa Fluor 488
Formule PBS with 5mM sodium azide; pH 7.5
Espèces hôtes Chicken
Immunogène Gamma Immunoglobins Heavy and Light chains.
Méthode de purification Purified
Quantité 1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage 4°C, store in dark
Type de produit Secondary Antibody
Forme Liquid
Isotype IgY
Afficher plus de résultats Afficher moins de résultats