missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Mouse IgG (H+L) Secondary Antibody

Catalog No. SA172018
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172018 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172018 Supplier Invitrogen™ Supplier No. SA172018
  • ONLINE EXCLUSIVE PRICE
    $358.64
    $208.50 / Each

    Save $150.14

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

SA1-72018 detects Mouse IgG (heavy and light chains) in mouse samples. It also cross-reacts with human IgG, rat IgG and porcine IgG, but has minimal reactivity with rabbit, goat, guinea pig, cow, ovine, or horse IgG. SA1-72018 has been successfully used in ELISA and Western blot procedures. The SA1-72018 immunogen is from purified mouse IgG.

Mouse IgG is an immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant immunoglobulin in the serum and plays a crucial role in the adaptive immune response. Mouse IgG is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. Mouse IgG is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.

Specifications

Antigen Mouse IgG (H+L)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.075% sodium azide; pH 7.2
Host Species Chicken
Immunogen Purified mouse IgG.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less