missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Mouse IgG (H+L) Secondary Antibody

Catalog No. A15977
Encompass
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15977 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15977 Supplier Invitrogen™ Supplier No. A15977

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Mouse IgG is an immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant immunoglobulin in the serum and plays a crucial role in the adaptive immune response. Mouse IgG is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. Mouse IgG is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.

Specifications

Antigen Mouse IgG (H+L)
Applications ELISA
Classification Polyclonal
Concentration >4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Host Species Chicken
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less