missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Human IgG (H+L) Secondary Antibody, FITC

Catalog No. SA172037
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172037 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172037 Supplier Invitrogen™ Supplier No. SA172037
  • ONLINE EXCLUSIVE PRICE
    $419.27
    $243.75 / Each

    Save $175.52

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

SA1-72037 detects Human IgG in human samples. SA1-72037 has been successfully used in immunohistochemistry procedures. The SA1-72037 immunogen is from purified normal human IgG.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgG (H+L)
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.075% sodium azide
Host Species Chicken
Immunogen Purified normal human IgG.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less