missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Human IgG F(ab')2 Secondary Antibody, Biotin

Catalog No. SA172044
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172044 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172044 Supplier Invitrogen™ Supplier No. SA172044

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

SA1-72044 detects the Fab'2 fragment of IgG in human samples. SA1-72044 has been successfully used in ELISA and western blot procedures. The SA1-72044 immunogen is from purified IgG from normal human serum.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgG F(ab')2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.1% chloroacetamide; pH 7.2
Host Species Chicken
Immunogen Purified IgG from normal human serum.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less