missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Bovine IgG (H+L) Secondary Antibody, Biotin

Catalog No. SA172014
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172014 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172014 Supplier Invitrogen™ Supplier No. SA172014
  • ONLINE EXCLUSIVE PRICE
    $406.38
    $236.25 / Each

    Save $170.13

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chicken Polyclonal Secondary Antibody

SA1-72014 detects Bovine IgG in bovine samples. SA1-72014 has been successfully used in ELISA procedures. The SA1-72014 immunogen is from purified bovine IgG isolated from normal pooled bovine serum.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Bovine IgG (H+L)
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.01% thimerosal
Host Species Chicken
Immunogen Purified bovine IgG isolated from normal pooled bovine serum.
Purification Method purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgY
Show More Show Less