missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Attached Hood, Elastic Wrists and Ankles
These durable and disposable coveralls with serged seams and elastic back protect against chemical splashes and toxic particles.
$126.74 - $156.46
Specifications
| Product Type | Chemical Protective Clothing |
|---|---|
| Gender | Unisex |
| Material | Laminated Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-184-126
|
International Enviroguard™
7015YSM |
Medium |
Case of 12 Each for $126.74
|
|
|||||
|
17-184-127
|
International Enviroguard™
7015YSL |
Large |
Case of 12 Each for $129.47
|
|
|||||
|
17-184-125
|
International Enviroguard™
7015YSXL |
X-Large |
Case of 12 Each for $135.91
|
|
|||||
|
17-184-131
|
International Enviroguard™
7015YS2XL |
2X-Large |
Case of 12 Each for $141.00
|
|
|||||
|
17-184-130
|
International Enviroguard™
7015YS3XL |
3X-Large |
Case of 12 Each for $146.65
|
|
|||||
|
17-184-129
|
International Enviroguard™
7015YS4XL |
4X-Large |
Case of 12 Each for $151.30
|
|
|||||
|
17-184-128
|
International Enviroguard™
7015YS5XL |
5X-Large |
Case of 12 Each for $156.46
|
|
|||||
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- Elastic wrists and ankles provide a secure closure that helps keep pants and sleeves in place to prevent skin exposure
- Roomy, seamless shoulder area resists tears to provide more protection in high-movement areas
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- High moisture vapor transfer rate (MVTR) allows heat and moisture vapor to escape, so heat isn't trapped near the body
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
| Chemical Protective Clothing | |
| Laminated Non-woven Fabric | |
| Single-use | |
| Elastic | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes |
| Unisex | |
| Yellow | |
| Elastic at Back | |
| Attached, Elastic Face | |
| Serged | |
| Chemicals, Acid, Caustics, Isopropyl Alcohol, Particle | |
| None |