missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Attached Hood, Elastic Wrists and Ankles
These durable and disposable coveralls with serged seams and elastic back protect against chemical splashes and toxic particles.
Supplier: International Enviroguard™ 7015YS4XL
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- Elastic wrists and ankles provide a secure closure that helps keep pants and sleeves in place to prevent skin exposure
- Roomy, seamless shoulder area resists tears to provide more protection in high-movement areas
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- High moisture vapor transfer rate (MVTR) allows heat and moisture vapor to escape, so heat isn't trapped near the body
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
| Chemical Protective Clothing | |
| Unisex | |
| Yellow | |
| Elastic at Back | |
| Elastic | |
| Attached, Elastic Face | |
| Serged | |
| Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle | |
| None |
| 4X-Large | |
| Laminated Non-woven Fabric | |
| Single-use | |
| Elastic | |
| Hooded | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Yes |